Bromuro de (1-heptil)trifenilfosfonio ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: Heptyltriphenylphosphonium bromide

H0545-25G H0545-500G
TCIAH0545-25GEA 38 EUR
TCIAH0545-25G TCIAH0545-500G
Bromuro de (1-heptil)trifenilfosfonio ≥98.0% (by HPLC, titration analysis)
Bromuro de (1-heptil)trifenilfosfonio
Formula: H₃C(CH₂)₆P(C₆H₅)₃Br
MW: 441.39 g/mol
Número MDL: MFCD00050249
Núm. CAS: 13423-48-8
EINECS: 236-539-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1e196b9df6d14d948b69059e2c604479f8f2a79&vid=cb2b0444fed0ac3b3663e33fa6a831e745810ed6&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR