Ácido (3,5-dimetoxifenil)acético ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
2-(3,5-Dimethoxyphenyl)acetic acid
D2950-1G
D2950-5G
TCIAD2950-1GEA
24
EUR
TCIAD2950-1G
TCIAD2950-5G
Ácido (3,5-dimetoxifenil)acético ≥98.0% (by titrimetric analysis)
Ácido (3,5-dimetoxifenil)acético
Formula:
(C₆H₃(OCH₃)₂)CH₂COOH |
Número MDL:
MFCD00016827 Núm. CAS: 4670-10-4 |
|
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 96.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 101.0 to 105.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...