Ácido (4-nitrofenil)acético ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
4-Nitrophenylacetic acid
N0227-25G
N0227-500G
TCIAN0227-25GEA
19
EUR
TCIAN0227-25G
TCIAN0227-500G
Ácido (4-nitrofenil)acético ≥98.0% (by HPLC, titration analysis)
Ácido (4-nitrofenil)acético
Formula:
(C₆H₄(NO₂))CH₂COOH MW: 181.15 g/mol |
Número MDL:
MFCD00007383 Núm. CAS: 104-03-0 EINECS: 203-168-5 Índice Merck: 14,06621 |
|
Resultados test especificaciones
Appearance | White to Green to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 153.0 to 157.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...