2-Amino-3-nitrofenol ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
2-Hydroxy-6-nitroaniline
A1425-25G
A1425-5G
TCIAA1425-25GEA
61
EUR
TCIAA1425-25G
TCIAA1425-5G
2-Amino-3-nitrofenol ≥98.0% (by HPLC, titration analysis)
2-Amino-3-nitrofenol
Formula:
C₆H₆N₂O₃ MW: 154.13 g/mol |
Número MDL:
MFCD00010875 Núm. CAS: 603-85-0 EINECS: 210-060-1 |
|
Resultados test especificaciones
Appearance | Orange to Brown to Dark purple powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...