3,5-Di-terc-butilanilina ≥98.0% (por GC)

Proveedor: TCI
Atención

D2515-1G D2515-5G
TCIAD2515-1GEA 82 EUR
TCIAD2515-1G TCIAD2515-5G
3,5-Di-terc-butilanilina ≥98.0% (por GC)
3,5-Di-terc-butilanilina
Formula: C₁₄H₂₃N
MW: 205,34 g/mol
Número MDL: MFCD00134096
Núm. CAS: 2380-36-1
EINECS: 219-173-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4be48c9fd339f938e15012abd33e906f484430d&vid=f4be48c9fd339f938e15012abd33e906f484430d&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 53.0 to 57.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR