2-Amino-5-cloropiridina ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
5-Chloropyridin-2-amine, 5-Chloro-2-pyridylamine
A0837-25G
A0837-500G
A0837-100G
TCIAA0837-25GEA
34
EUR
TCIAA0837-25G
TCIAA0837-500G
TCIAA0837-100G
2-Amino-5-cloropiridina ≥98.0% (by GC, titration analysis)
2-Amino-5-cloropiridina
Formula:
C₅H₅ClN₂ MW: 128.56 g/mol Pto. Ebullición: 241 |
Número MDL:
MFCD00006324 Núm. CAS: 1072-98-6 EINECS: 214-020-4 |
|
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 135.0 to 138.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...