3-Fluorobenzamida ≥97.0% (by HPLC, total nitrogen)
Proveedor: TCI
F0266-25G
F0266-5G
TCIAF0266-25GEA
249
EUR
TCIAF0266-25G
TCIAF0266-5G
3-Fluorobenzamida ≥97.0% (by HPLC, total nitrogen)
3-Fluorobenzamida
Formula:
C₇H₆FNO MW: 139,13 g/mol Pto. Ebullición: 287 °C (1013 hPa) Pto. de fusión: 133 °C |
Número MDL:
MFCD00007983 Núm. CAS: 455-37-8 EINECS: 207-247-5 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Melting point | 130.0 to 132.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...