4-Amino-2,6-dicloropiridina ≥97.0% (by GC, total nitrogen)
Proveedor: TCI
|
Atención
|
Sinónimos:
2,6-Dichloro-4-pyridylamine, 2,6-Dichloropyridin-4-amine
A2369-1G
A2369-5G
TCIAA2369-1GEA
19
EUR
TCIAA2369-1G
TCIAA2369-5G
4-Amino-2,6-dicloropiridina ≥97.0% (by GC, total nitrogen)
4-Amino-2,6-dichloropyridine
Formula:
C₅H₄Cl₂N₂ MW: 163,01 g/mol |
Número MDL:
MFCD00052832 Núm. CAS: 2587-02-2 |
Resultados test especificaciones
Appearance | White to Amber powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (with Total Nitrogen) | min. 97.0 % |
Melting point | 173.0 to 177.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...