trans-2,6-Difluorocinnamic acid ≥95.0% (por GC)
Proveedor: TCI
|
Atención
|
Sinónimos:
(E)-3-(2,6-Difluorophenyl)acrylic acid
D4115-1G
D4115-5G
TCIAD4115-1GEA
20
EUR
TCIAD4115-1G
TCIAD4115-5G
trans-2,6-Difluorocinnamic acid ≥95.0% (por GC)
trans-2,6-Difluorocinnamic acid
Formula:
C₉H₆F₂O₂ MW: 184,14 g/mol |
Número MDL:
MFCD00010319 Núm. CAS: 102082-89-3 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 121.0 to 125.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...