D(-)-Norvalina ≥99.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: (R)-(-)-Norvalina, H-D-Nva-OH

N0673-1G N0673-25G N0673-5G
TCIAN0673-1GEA 74 EUR
TCIAN0673-1G TCIAN0673-25G TCIAN0673-5G
D(-)-Norvalina ≥99.0% (by titrimetric analysis)
D(-)-Norvalina
Formula: C₅H₁₁NO₂
MW: 117,15 g/mol
Número MDL: MFCD00008097
Núm. CAS: 2013-12-9
EINECS: 217-936-2
Índice Merck: 14,06716
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5287778bf6d7853c97668782aecbb0aaa191b289&vid=5287778bf6d7853c97668782aecbb0aaa191b289&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Specific rotation [a]20/D -24.0 to -26.0 deg (C=10, 6mol/L HCl)
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR