5-Cloro-1,10-fenantrolina ≥98.0% (by titrimetric analysis)
Proveedor: TCI
C0786-1G
C0786-100MG
TCIAC0786-1GEA
116
EUR
TCIAC0786-1G
TCIAC0786-100MG
5-Cloro-1,10-fenantrolina ≥98.0% (by titrimetric analysis)
5-Cloro-1,10-fenantrolina
Formula:
C₁₂H₇ClN₂ |
Número MDL:
MFCD00004980 Núm. CAS: 4199-89-7 EINECS: 224-098-1 |
|
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 123.0 to 126.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...