m-Anísohidrazida ≥98.0% (by GC, titration analysis)
Proveedor: TCI
Sinónimos:
3-Metoxibenzohidrazida, 3-Anísohidrazida
A1446-25G
A1446-5G
TCIAA1446-25GEA
171
EUR
TCIAA1446-25G
TCIAA1446-5G
m-Anísohidrazida ≥98.0% (by GC, titration analysis)
m-Anísohidrazida
Formula:
C₈H₁₀N₂O₂ MW: 166,18 g/mol |
Número MDL:
MFCD00007601 Núm. CAS: 5785-06-8 EINECS: 227-310-0 |
Resultados test especificaciones
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...