Ácido 2,4,5-trifluoro-3-metoxibenzoico ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: Moxidectin Impurity 5, 3-Methoxy-2,4,5-trifluorobenzoic acid

T1917-25G T1917-5G
TCIAT1917-25GEA 52 EUR
TCIAT1917-25G TCIAT1917-5G
Ácido 2,4,5-trifluoro-3-metoxibenzoico ≥98.0% (by GC, titration analysis)
Ácido 2,4,5-trifluoro-3-metoxibenzoico
Formula: C₈H₅F₃O₃
MW: 206,12 g/mol
Número MDL: MFCD00153201
Núm. CAS: 112811-65-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8fd0fcef9ff0b2943b0e920249ca97bb3fea78e1&vid=8fd0fcef9ff0b2943b0e920249ca97bb3fea78e1&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 116.0 to 119.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR