Ácido 2,3,5,6-tetrafluoro-4-hidroxibenzoico ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

T1943-1G T1943-5G
TCIAT1943-1GEA 114 EUR
TCIAT1943-1G TCIAT1943-5G
Ácido 2,3,5,6-tetrafluoro-4-hidroxibenzoico ≥98.0% (by titrimetric analysis)
Ácido 2,3,5,6-tetrafluoro-4-hidroxibenzoico
Formula: C₇H₂F₄O₃
MW: 210,08 g/mol
Número MDL: MFCD00129952
Núm. CAS: 652-34-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d521f4da23743e86b66327294fa20d034cab976&vid=3d521f4da23743e86b66327294fa20d034cab976&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 152.0 to 156.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR