α-Bromo-3-cloro-2-fluorotolueno ≥96.0% (by GC, titration analysis)
Proveedor: TCI
|
Peligro
|
C2118-1G
C2118-5G
TCIAC2118-1GEA
35
EUR
TCIAC2118-1G
TCIAC2118-5G
α-Bromo-3-cloro-2-fluorotolueno ≥96.0% (by GC, titration analysis)
α-Bromo-3-cloro-2-fluorotolueno
Formula:
C₇H₅BrClF Pto. Ebullición: 125 - 126 °C (15 -rr) |
Número MDL:
MFCD00042503 Núm. CAS: 85070-47-9 UN: 3261 ADR: 8,II |
|
Resultados test especificaciones
Appearance | White to Orange to Green powder to lump |
Purity (GC) | min. 96.0 % |
Purity (Argentmetric Titration) | min. 96.0 % |
Freezing point | 27.0 to 32.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...