α-Bromo-4-cloro-3-fluorotolueno ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Peligro

C2484-1G C2484-5G
TCIAC2484-1GEA 29 EUR
TCIAC2484-1G TCIAC2484-5G
α-Bromo-4-cloro-3-fluorotolueno ≥98.0% (by GC, titration analysis)
α-Bromo-4-cloro-3-fluorotolueno
Formula: C₇H₅BrClF
Número MDL: MFCD04115859
Núm. CAS: 206362-80-3
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=679748897e9b0c9de237cce535fe3980e8c94987&vid=679748897e9b0c9de237cce535fe3980e8c94987&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to lump
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR