Alcohol 2-cloro-4-fluorobencílico ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
C2527-25G
C2527-5G
TCIAC2527-25GEA
258
EUR
TCIAC2527-25G
TCIAC2527-5G
Alcohol 2-cloro-4-fluorobencílico ≥98.0% (por GC)
Alcohol 2-cloro-4-fluorobencílico
Formula:
C₇H₆ClFO |
Número MDL:
MFCD00236024 Núm. CAS: 208186-84-9 |
|
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 45.0 to 49.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...