4,4'-Diiodobifenilo ≥98.0% (por GC)

Proveedor: TCI

Sinónimos: 4,4'-Diiodo-1,1'-biphenyl

D2315-25G D2315-250G
TCIAD2315-25GEA 25 EUR
TCIAD2315-25G TCIAD2315-250G
4,4'-Diiodobifenilo ≥98.0% (por GC)
4,4'-Diiodobifenilo
Formula: C₁₂H₈I₂
MW: 406 g/mol
Número MDL: MFCD00001057
Núm. CAS: 3001-15-8
EINECS: 221-080-5
UN: 3152
ADR: 9,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42a9410f18f4580b303e4d8f0e2cdc1f9eff38e7&vid=42a9410f18f4580b303e4d8f0e2cdc1f9eff38e7&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 206.0 to 209.0 deg-C
Solubility in hot Toluene very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR