Acetoacetato de litio ≥97.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
3-Oxobutyric acid Lithium salt, Lithium 3-Oxobutyrate, Acetoacetic acid Lithium salt
A1478-1G
A1478-5G
TCIAA1478-1GEA
111
EUR
TCIAA1478-1G
TCIAA1478-5G
Acetoacetato de litio ≥97.0% (by titrimetric analysis)
Acetoacetato de litio
Formula:
C₄H₅LiO₃ MW: 108,02 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD00013060 Núm. CAS: 3483-11-2 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 97.0 % |
Solubility in Water | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...