4-Bromo-2-iodoanilina ≥97.0% (by GC, titration analysis)

Proveedor: TCI
Peligro

B3164-1G B3164-5G
TCIAB3164-1GEA 29 EUR
TCIAB3164-1G TCIAB3164-5G
4-Bromo-2-iodoanilina ≥97.0% (by GC, titration analysis)
4-Bromo-2-iodoanilina
Formula: C₆H₅BrIN
Número MDL: MFCD02093764
Núm. CAS: 66416-72-6
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78cf5f3186260da00c85c735e6ff80dc11302d96&vid=78cf5f3186260da00c85c735e6ff80dc11302d96&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Gray to Red powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point gt. 68.0 to 72.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR