2-Hidroxi-1-naftaldehído ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
2-Hydroxy-α-naphthaldehyde
H0275-25G
H0275-100G
TCIAH0275-25GEA
66
EUR
TCIAH0275-25G
TCIAH0275-100G
2-Hidroxi-1-naftaldehído ≥98.0% (by GC, titration analysis)
2-Hidroxi-1-naftaldehído
Formula:
C₁₁H₈O₂ MW: 172,18 g/mol Pto. Ebullición: 192°C |
Número MDL:
MFCD00004005 Núm. CAS: 708-06-5 EINECS: 211-902-0 |
Resultados test especificaciones
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 81.0 to 84.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...