L-Homoserina ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: (S)-(-)-Homoserina, H-Hser-OH

H1030-1G H1030-5G
TCIAH1030-1GEA 33 EUR
TCIAH1030-1G TCIAH1030-5G
L-Homoserina ≥98.0% (by titrimetric analysis)
L-Homoserina
Formula: C₄H₉NO₃
Pto. de fusión: 203 °C
Número MDL: MFCD00063090
Núm. CAS: 672-15-1
EINECS: 211-590-6
Índice Merck: 14,04739
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b3a4d6a67206897dc4c0ffac2dc52bc1624712e&vid=5b3a4d6a67206897dc4c0ffac2dc52bc1624712e&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -6.0 to -10.0 deg (C=2,H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR