Ácido (4-hidroxifenoxi)acético ≥99.0% (by titrimetric analysis)

Proveedor: TCI

H0992-25G H0992-5G
TCIAH0992-25GEA 214 EUR
TCIAH0992-25G TCIAH0992-5G
Ácido (4-hidroxifenoxi)acético ≥99.0% (by titrimetric analysis)
Ácido (4-hidroxifenoxi)acético
Formula: (C₆H₄(OH))OCH₂COOH
MW: 168.15 g/mol
Número MDL: MFCD00014362
Núm. CAS: 1878-84-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27ed5929a7ba946ca9057787fed3c4af31b97e19&vid=27ed5929a7ba946ca9057787fed3c4af31b97e19&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 99.0 %
Melting point 154.0 to 157.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR