Hexafluorofosfato de 1-butil-2,3-dimetilimidazolio ≥97.0% (by total nitrogen and titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
1-Butyl-2,3-dimethyl-1H-imidazol-3-ium hexafluorophosphate(V)
B2474-25G
B2474-5G
TCIAB2474-25GEA
267
EUR
TCIAB2474-25G
TCIAB2474-5G
Hexafluorofosfato de 1-butil-2,3-dimetilimidazolio ≥97.0% (by total nitrogen and titration analysis)
Hexafluorofosfato de 1-butil-2,3-dimetilimidazolio
Formula:
C₉H₁₇F₆N₂P MW: 298,21 g/mol |
Número MDL:
MFCD03790877 Núm. CAS: 227617-70-1 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (with Total Nitrogen) | min. 97.0 % |
Purity (Ion exchange titration) | min. 97.0 % |
Melting point | 36.0 to 42.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...