2,6-Dihidroxibenzoato de metilo ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Methyl 2,6-dihydroxybenzoate, 2,6-Dihydroxybenzoic acid methyll ester, gamma-Resorcylic acid methyll ester
D1461-25G
D1461-5G
TCIAD1461-25GEA
228
EUR
TCIAD1461-25G
TCIAD1461-5G
2,6-Dihidroxibenzoato de metilo ≥98.0% (by GC, titration analysis)
2,6-Dihidroxibenzoato de metilo
Formula:
C₈H₈O₄ MW: 168,15 g/mol |
Número MDL:
MFCD00059619 Núm. CAS: 2150-45-0 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 68.0 to 72.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...