3,5-Dimetil-4-iodoisoxazol ≥98.0% (por GC)

Proveedor: TCI

I0727-1G I0727-5G
TCIAI0727-1GEA 29 EUR
TCIAI0727-1G TCIAI0727-5G
3,5-Dimetil-4-iodoisoxazol ≥98.0% (por GC)
3,5-Dimetil-4-iodoisoxazol
Formula: C₅H₆INO
MW: 223,01 g/mol
Número MDL: MFCD00173743
Núm. CAS: 10557-85-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6048d2baec992bea644523236428b0ac539ba0e6&vid=6048d2baec992bea644523236428b0ac539ba0e6&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 52.0 to 56.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR