5-Amino-3-fenilisoxazol ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI
Atención

A2437-1G A2437-5G
TCIAA2437-1GEA 58 EUR
TCIAA2437-1G TCIAA2437-5G
5-Amino-3-fenilisoxazol ≥98.0% (by HPLC, total nitrogen)
5-Amino-3-phenylisoxazole
Formula: C₉H₇NO
MW: 145,16 g/mol
Número MDL: MFCD00464114
Núm. CAS: 4369-55-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab709fd3eb86e8d15f53cd07b257b7e9896cd170&vid=ab709fd3eb86e8d15f53cd07b257b7e9896cd170&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 111.0 to 115.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR