N-Ciclohexilmaleimida ≥98.0% (por GC)
Proveedor: TCI
|
Peligro
|
C1044-25G
C1044-100G
TCIAC1044-25GEA
69
EUR
TCIAC1044-25G
TCIAC1044-100G
N-Ciclohexilmaleimida ≥98.0% (por GC)
N-Ciclohexilmaleimida
Formula:
C₁₀H₁₃NO₂ MW: 179,22 g/mol Pto. Ebullición: 132°C |
Número MDL:
MFCD00043904 Núm. CAS: 1631-25-0 EINECS: 216-630-6 UN: 2811 ADR: 6.1,II |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 87.0 to 91.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...