2,2'-Ditiodianilina ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Peligro
|
Sinónimos:
Disulfuro de bis(2-aminofenilo)
D1246-25G
D1246-500G
TCIAD1246-25GEA
98
EUR
TCIAD1246-25G
TCIAD1246-500G
2,2'-Ditiodianilina ≥98.0% (by HPLC, titration analysis)
2,2'-Ditiodianilina
Formula:
C₁₂H₁₂N₂S₂ MW: 248.37 g/mol |
Número MDL:
MFCD00007703 Núm. CAS: 1141-88-4 EINECS: 214-529-1 |
|
Resultados test especificaciones
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 91.0 to 95.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...