4-Amino-3-nitrobenzonitrilo ≥98.0% (por GC)
Proveedor: TCI
|
Peligro
|
A2447-25G
A2447-5G
TCIAA2447-25GEA
456
EUR
TCIAA2447-25G
TCIAA2447-5G
4-Amino-3-nitrobenzonitrilo ≥98.0% (por GC)
4-Amino-3-nitrobenzonitrile
Formula:
C₇H₅N₃O₂ MW: 163,14 g/mol |
Número MDL:
MFCD00013373 Núm. CAS: 6393-40-4 UN: 3439 ADR: 6.1,III |
Resultados test especificaciones
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 160.0 to 164.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...