Anaranjado de xilenol, sal disódica
Proveedor: TCI
Sinónimos:
3,3′-Bis[N,N-bis(carboxymethyl)aminomethyl]-o-cresolsulphonephthalein disodium salt
B0477-1G
B0477-25G
B0477-5G
TCIAB0477-1GEA
21
EUR
TCIAB0477-1G
TCIAB0477-25G
TCIAB0477-5G
Anaranjado de xilenol, sal disódica
Anaranjado de xilenol, sal disódica
Formula:
C₃₁H₃₀N₂Na₂O₁₃S |
Número MDL:
MFCD00014580 Núm. CAS: 1202864-39-8 EINECS: 216-553-8 |
|
Resultados test especificaciones
Appearance | Orange to Brown to Dark red powder to crystal |
Lambda max. | 433.0 to 437.0 nm (H2O) |
Absorbance | min. 0.210 (0.01g/L,H2O,433.0 to 437.0 nm) (after drying) |
Sensitiveness | Coloring by Zinc solution. Fading by EDTA2Na. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...