5,6-Dimetil-1,10-fenantrolina ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
D1559-1G
D1559-100MG
TCIAD1559-1GEA
351
EUR
TCIAD1559-1G
TCIAD1559-100MG
5,6-Dimetil-1,10-fenantrolina ≥98.0% (by HPLC, titration analysis)
5,6-Dimetil-1,10-fenantrolina
Formula:
C₁₄H₁₂N₂ MW: 208,26 g/mol |
Número MDL:
MFCD00004983 Núm. CAS: 3002-81-1 EINECS: 221-100-2 |
Resultados test especificaciones
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...