5-Bromo-3-nitrosalicilaldehído ≥97.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
5-Bromo-2-hidroxi-3-nitrobenzaldehído
B0908-1G
B0908-5G
TCIAB0908-1GEA
31
EUR
TCIAB0908-1G
TCIAB0908-5G
5-Bromo-3-nitrosalicilaldehído ≥97.0% (by GC, titration analysis)
5-Bromo-3-nitrosalicilaldehído
Formula:
C₇H₄BrNO₄ MW: 246.02 g/mol |
Número MDL:
MFCD00130110 Núm. CAS: 16634-88-1 |
|
Resultados test especificaciones
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 123.0 to 127.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...