6-Bromo-3-metoxisalicilaldehído ≥99.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
6-Bromo-2-hidroxi-3-metoxibenzaldehído
B1487-1G
B1487-25G
B1487-5G
TCIAB1487-1GEA
27
EUR
TCIAB1487-1G
TCIAB1487-25G
TCIAB1487-5G
6-Bromo-3-metoxisalicilaldehído ≥99.0% (by GC, titration analysis)
6-Bromo-3-metoxisalicilaldehído
Formula:
C₈H₇BrO₃ MW: 231,05 g/mol |
Número MDL:
MFCD00046152 Núm. CAS: 20035-41-0 |
Resultados test especificaciones
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 99.0 % |
Purity (Neutralization titration) | min. 99.0 % |
Melting point | 100.0 to 104.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...