4-Bromosalicilaldehído ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 4-Bromo-2-hidroxibenzaldehído

B3637-1G B3637-5G
TCIAB3637-1GEA 83 EUR
TCIAB3637-1G TCIAB3637-5G
4-Bromosalicilaldehído ≥98.0% (by GC, titration analysis)
4-Bromosalicilaldehído
Formula: C₇H₅BrO₂
MW: 201,02 g/mol
Número MDL: MFCD06252606
Núm. CAS: 22532-62-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8dad4e4ad1c101e877ff084182fcd9ae8e5a7f0e&vid=8dad4e4ad1c101e877ff084182fcd9ae8e5a7f0e&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 51.0 to 55.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR