4-Hidroxibenzamida ≥98.0% (by total nitrogen basis)
Proveedor: TCI
|
Atención
|
H0200-25G
H0200-5G
TCIAH0200-25GEA
93
EUR
TCIAH0200-25G
TCIAH0200-5G
4-Hidroxibenzamida ≥98.0% (by total nitrogen basis)
4-Hidroxibenzamida
Formula:
C₇H₇NO₂ |
Número MDL:
MFCD00007997 Núm. CAS: 619-57-8 EINECS: 210-602-7 |
|
Resultados test especificaciones
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 159.0 to 163.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...