1-Butilpiridinio hexafluorofosfato ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Hexafluorofosfato de 1-butilpiridinio, Hexafluorofosfato de butilpiridinio
B2196-25G
B2196-5G
TCIAB2196-25GEA
95
EUR
TCIAB2196-25G
TCIAB2196-5G
1-Butilpiridinio hexafluorofosfato ≥98.0% (by HPLC, titration analysis)
1-Butilpiridinio hexafluorofosfato
Formula:
C₉H₁₄F₆NP MW: 281.18 g/mol |
Número MDL:
MFCD03093297 Núm. CAS: 186088-50-6 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Ion exchange titration) | min. 98.0 % |
Melting point | 73.0 to 77.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...