2-Isopropilimidazol ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 2-Isopropil-1H-imidazol

I0313-25G I0313-500G
TCIAI0313-25GEA 16 EUR
TCIAI0313-25G TCIAI0313-500G
2-Isopropilimidazol ≥98.0% (by GC, titration analysis)
2-Isopropilimidazol
Formula: C₆H₁₀N₂
MW: 110,16 g/mol
Número MDL: MFCD00014486
Núm. CAS: 36947-68-9
EINECS: 253-286-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e44f7dab75562a3e67ce976631d02e8f65e69cd0&vid=e44f7dab75562a3e67ce976631d02e8f65e69cd0&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 129.0 to 135.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR