Potasio (bromometil)trifluoroborato ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: (Bromomethyl)trifluoroboric acid potassium salt, Potassium (bromomethyl)trifluoroborate

P1807-1G P1807-5G
TCIAP1807-1GEA 34 EUR
TCIAP1807-1G TCIAP1807-5G
Potasio (bromometil)trifluoroborato ≥98.0% (by titrimetric analysis)
Potasio (bromometil)trifluoroborato
Formula: CH₂BBrF₃K
MW: 200.83 g/mol
Pto. de fusión: 220°C
Número MDL: MFCD09265154
Núm. CAS: 888711-44-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efe35f3988da87f915267828ee1207ce86d2c5ec&vid=efe35f3988da87f915267828ee1207ce86d2c5ec&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR