Potasio feniltrifluoroborato ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: Potassium trifluoro(phenyl)borate, Phenyltrifluoroboric acid potassium salt

P1582-1G P1582-25G P1582-5G
TCIAP1582-1GEA 35 EUR
TCIAP1582-1G TCIAP1582-25G TCIAP1582-5G
Potasio feniltrifluoroborato ≥98.0% (by HPLC, titration analysis)
Potasio feniltrifluoroborato
Formula: C₆H₅BF₃K
MW: 184,01 g/mol
Número MDL: MFCD01318172
Núm. CAS: 153766-81-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48f8cfbe605cb85dfdf6e0e3b71b9a7483f31064&vid=48f8cfbe605cb85dfdf6e0e3b71b9a7483f31064&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR