2-Metil-2-propilpropano-1,3-diol ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
Sinónimos:
2,2-Bis(hydroxymethyl)pentane, 2-Methyl-2-propylpropane-1,3-diol, 2,2-Dimethylolpentane
D0766-25G
D0766-500G
TCIAD0766-25GEA
16
EUR
TCIAD0766-25G
TCIAD0766-500G
2-Metil-2-propilpropano-1,3-diol ≥98.0% (por GC)
2-Metil-2-propilpropano-1,3-diol
Formula:
C₇H₁₆O₂ |
Número MDL:
MFCD00004696 Núm. CAS: 78-26-2 EINECS: 201-099-5 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 56.0 to 60.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...