(S)-(-)-3-Amino-1,2-propanodiol ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Peligro

A1980-1G A1980-5G
TCIAA1980-1GEA 30 EUR
TCIAA1980-1G TCIAA1980-5G
(S)-(-)-3-Amino-1,2-propanodiol ≥98.0% (by GC, titration analysis)
(S)-(-)-3-Amino-1,2-propanodiol
Formula: C₃H₉NO₂
Pto. Ebullición: 130°C
Número MDL: MFCD00798260
Núm. CAS: 61278-21-5
UN: 3259
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36d33d3f1233ac51629e19fc1d3791659c300886&vid=36d33d3f1233ac51629e19fc1d3791659c300886&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 54.0 to 60.0 deg-C
Specific rotation [a]20/D -2.0 to -4.0 deg (C=5, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR