[(Fenilmetil)sulfonil]benceno ≥98.0% (por GC)

Proveedor: TCI

Sinónimos: ((Phenylmethyl)sulphonyl)benzol, [(Phenylmethyl)sulphonyl]benzene, Benzylsulphonylbenzene, (Benzylsulphonyl)benzene

B3080-25G B3080-5G
TCIAB3080-25GEA 196 EUR
TCIAB3080-25G TCIAB3080-5G
[(Fenilmetil)sulfonil]benceno ≥98.0% (por GC)
[(Fenilmetil)sulfonil]benceno
Formula: C₁₃H₁₂O₂S
Número MDL: MFCD00025040
Núm. CAS: 3112-88-7
EINECS: 221-477-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d209c903db5d54b73b64341dbdaf8e2bc76e4595&vid=d209c903db5d54b73b64341dbdaf8e2bc76e4595&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 146.0 to 150.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR