4-Amino-2-metoxibenzoato de metilo ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
M1998-25G
M1998-5G
TCIAM1998-25GEA
74
EUR
TCIAM1998-25G
TCIAM1998-5G
4-Amino-2-metoxibenzoato de metilo ≥98.0% (by GC, titration analysis)
4-Amino-2-metoxibenzoato de metilo
Formula:
C₉H₁₁NO₃ |
Número MDL:
MFCD00017202 Núm. CAS: 27492-84-8 EINECS: 248-494-9 |
|
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 157.0 to 161.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...