Ácido 5-bromo-3,5-dihidroxibenzoico ≥97.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
B1425-25G
B1425-250G
TCIAB1425-25GEA
44
EUR
TCIAB1425-25G
TCIAB1425-250G
Ácido 5-bromo-3,5-dihidroxibenzoico ≥97.0% (by HPLC, titration analysis)
Ácido 5-bromo-3,5-dihidroxibenzoico
Formula:
C₇H₅BrO₄ |
Número MDL:
MFCD00002513 Núm. CAS: 16534-12-6 EINECS: 240-606-4 |
|
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...