Bis(4-fluorofenil) sulfona ≥99.0% (por GC)

Proveedor: TCI

D0537-10G D0537-25G D0537-250G
TCIAD0537-10GEA 52 EUR
TCIAD0537-10G TCIAD0537-25G TCIAD0537-250G
Bis(4-fluorofenil) sulfona ≥99.0% (por GC)
Bis(4-fluorofenil) sulfona
Formula: C₁₂H₈F₂O₂S
Número MDL: MFCD00000350
Núm. CAS: 383-29-9
EINECS: 206-847-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c338efa84a5baa21f05019db50c797ed7076b4e&vid=0c338efa84a5baa21f05019db50c797ed7076b4e&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Melting point 98.0 to 101.0 deg-C
Solubility in Methanol very faint turbidity

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR