Cloruro de metilamonio ≥98.0% (by total nitrogen basis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Metilamina clorhidrato
M0138-25G
M0138-500G
TCIAM0138-25GEA
16
EUR
TCIAM0138-25G
TCIAM0138-500G
Cloruro de metilamonio ≥98.0% (by total nitrogen basis)
Cloruro de metilamonio
Formula:
CH₆ClN MW: 67.52 g/mol |
Número MDL:
MFCD00012849 Núm. CAS: 593-51-1 EINECS: 209-795-0 |
|
Resultados test especificaciones
Appearance | White powder to crystaline |
Purity (Argentmetric Titration) | min. 98.0 % (calcd.on anh.substance) |
Purity (with Total Nitrogen) | min. 98.0 % (calcd.on anh.substance) |
Melting point | 226.0 to 236.0 deg-C |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...