2,4-Diamino-6-fenil-1,3,5-triazina ≥99.0% (by HPLC)
Proveedor: TCI
|
Atención
|
Sinónimos:
2,4-Diamino-6-fenil-s-triazina, 6-Fenil-1,3,5-triazina-2,4-diamina, 6-Fenil-1,3,5-triazin-2,4-diildiamina
D0111-25G
D0111-500G
TCIAD0111-25GEA
18
EUR
TCIAD0111-25G
TCIAD0111-500G
2,4-Diamino-6-fenil-1,3,5-triazina ≥99.0% (by HPLC)
2,4-Diamino-6-fenil-1,3,5-triazina
Formula:
C₉H₉N₅ MW: 187,2 g/mol |
Número MDL:
MFCD00023187 Núm. CAS: 91-76-9 EINECS: 202-095-6 Índice Merck: 14,01089 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 225.0 to 230.0 deg-C |
Solubility in N,N-DMF | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...