4-Bromofenilhidracinio cloruro ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
4-Bromofenilhidrazina clorhidrato
B0635-25G
B0635-5G
TCIAB0635-25GEA
118
EUR
TCIAB0635-25G
TCIAB0635-5G
4-Bromofenilhidracinio cloruro ≥98.0% (by HPLC, titration analysis)
4-Bromofenilhidracinio cloruro
Formula:
C₆H₈BrClN₂ MW: 223.50 g/mol |
Número MDL:
MFCD00012941 Núm. CAS: 622-88-8 |
|
Resultados test especificaciones
Appearance | White to Light yellow to Light red powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Potassium iodate Method) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...