4-Nitrofenilhidrazina clorhidrato ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 4-Nitrofenilhidracinio cloruro

N0233-25G N0233-5G
TCIAN0233-25GEA 129 EUR
TCIAN0233-25G TCIAN0233-5G
4-Nitrofenilhidrazina clorhidrato ≥98.0% (by HPLC, titration analysis)
4-Nitrofenilhidrazina clorhidrato
Formula: C₆H₇N₃O₂·HCl
MW: 189,6 g/mol
Número MDL: MFCD00040592
Núm. CAS: 636-99-7
EINECS: 211-273-2
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c774df6218013d6a12c514f73a38bd35122263e&vid=0c774df6218013d6a12c514f73a38bd35122263e&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Potassium iodate Method) min. 98.0 %
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR